Universal sorbent pads
Fournisseur: Brady
813809
BRDY813809EA 122.77 EUR InStock
BRDY813809
Universal sorbent pads
Matériaux absorbants
These grey coloured heavy weight, bonded and perforated universal sorbent pad absorbs industrial liquids including oils, water, solvents, coolants.
  • One sided spunbound for extra strength and less lint
  • Made of two-ply, spunbound/meltblown, bonded
Perforated to help minimise waste and lower disposal costs. Ideal sorbent for dealing with leaks and drips in the workplace and also suited for general industrial maintenance and spill clean up.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité