Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M7 Printers
Fournisseur: Brady
M7-17-498
BRDYM7-17-498EA 76.3 EUR InStock
BRDYM7-17-498
Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M7 Printers
Étiquettes
Permanent or repositionable, once applied the B-498 CleanLift® Series Vinyl Cloth remains in place, until it must be changed.
  • Adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • Semi-gloss finish for crisp, clean look
Labels are ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, generalMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité