BROEN-LAB UniFlex™ Flexible Stainless Steel Pipes for Special Water
Fournisseur: BROEN-LAB
93Z40401000
BROE93Z40401000EA 84.1 EUR InStock
BROE93Z40401000
BROEN-LAB UniFlex™ Flexible Stainless Steel Pipes for Special Water
Raccords et connecteurs
BROEN-LAB offers BROEN-LAB UniFlex™ hoses and flexible Stainless Steel pipes as an easy way to make the installation in the laboratory where safety and efficiency are required. BROEN-LAB UniFlex™ hoses are the ideal solution where there is a higher requirement for flexibility. Installation with flexible hoses saves time and labour - in other words - reduced installation costs.
  • Inner hose material: Stainless steel 1,4404 similar to AISI 316 L
  • Braiding: None
  • Materials with media contact: Stainless steel 1.4404, EPDM, aramide fibre/NBR gasket
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité