These TZe labelling tapes are developed for use with P-Touch labelprinters. They feature the exclusive laminated tape process and standard adhesive. Best suited for smooth flat surfaces.
- Perfect for everyday applications
- Stay on under normal indoor and outdoor use, including hot and cold environments
Length: 8,0 m
Different colours available.