Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-TrkA Rabbit Polyclonal Antibody

Fournisseur: BosterBio
PA1321
BSBTPA1321EA 0 EUR InStock
BSBTPA1321
Anti-TrkA Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for High affinity nerve growth factor receptor(NTRK1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Trk A(Neurotrophic tyrosine kinase receptor A) is the high affinity catalytic receptor for the neurotrophin, Nerve Growth Factor(NGF). Higher affinity binding of NGFR can achieved by association with higher molecular mass, low-affinity neurotrophin receptors, namely the tropomyosin receptor kinases, TRKA(NTRK1), TRKB(NTRK2), and TRKC(NTRK3). TRKA, TRKB, and TRKC are specific for or "preferred by" NGF, NTF4(Neurotrophin-4) and BDNF, and NTF3(Neurotrophin-3), respectively. NTF3 also binds to TRKA and TRKB, but with significantly lower affinity. The absence of TrkA(NGFR) expression was associated with a strong increase in the Sp3 repressor short isoform(s) and a lack of the Sp3 activator long isoform. Sp3 is a bifunctional transcription factor that has been reported to stimulate or repress the transcription of numerous genes. Indo et al.(1996) concluded that defects in TRKA cause CIPA(Congenital insensitivity to pain with anhidrosis) and that the NGF-TRKA system has a crucial role in the development and function of the nociceptive reception as well as establishment of thermoregulation via sweating in humans. These results also implicate genes encoding other TRK and neurotrophin family members as candidates for developmental defect(s) of the nervous system.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: TrkA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité