Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-Muscarinic Acetylcholine Receptor 2 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
PA1325-1
BSBTPA1325-1EA 552 EUR InStock
BSBTPA1325-1
Anti-Muscarinic Acetylcholine Receptor 2 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Muscarinic acetylcholine receptor M2(CHRM2) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

The muscarinic acetylcholine receptor M2, also known as the cholinergic receptor, muscarinic 2, is a muscarinic acetylcholine receptor. The M2 muscarinic receptors are located in the heart, where they act to slow the heart rate down to normal sinus rhythm after stimulatory actions of the sympathetic nervous system, by slowing the speed of depolarization. The CHRM2 gene inhibits the release of acetylcholine from cholinergic fibers in the lungs and elsewhere. In airway parasympathetic neurons, it is decreased by viral infections and by interferon-gamma, increasing actylcholine release. This gene is though to be involved in neuronal excitability, synaptic plasticity and feedback regulation of acetylcholine release and has previously been implicated in higher cognitive processing. In a sample of 667 individuals from 304 families, Gosso MF et al. genotyped three single-nucleotide polymorphisms(SNPs) in the CHRM2 gene on 7q31-35. CHRM2 is implicated in memory and cognition, functions impaired in many neuropsychiatric disorders. Wang et al. evidence of common and specific genetic effects: association of the muscarinic acetylcholine receptor M2(CHRM2) gene with alcohol dependence and major depressive syndrome.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

WB: The detection limit for CHRM2 is approximately 1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: Muscarinic Acetylcholine Receptor 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité