Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-FGF21 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
PA1673
BSBTPA1673EA 552 EUR InStock
BSBTPA1673
Anti-FGF21 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Fibroblast growth factor 21(FGF21) detection. Tested with WB in Human;Mouse;Rat.

FGF21(Fibroblast growth factor 21) is a protein that in humans is encoded by the FGF21 gene, which is also a member of the fibroblast growth factor(FGF) family. The FGF21 gene is mapped on 19q13.33. Using RT-PCR, Fgf21 was expressed in several types of adipose tissue in mice, including subcutaneous and epididymal fat pads and brown adipose tissue. The level of Fgf21 expression in adipose tissue was comparable to that in liver. FGF21 stimulates glucose uptake in adipocytes but not in other cell types. This effect is additive to the activity of insulin. FGF21 treatment of adipocytes is associated with phosphorylation of FRS2, a protein linking FGF receptors to the Ras/MAP kinase pathway. FGF21 also protects animals from diet-induced obesity when overexpressed in transgenic mice and lowers blood glucose and triglyceride levels when administered to diabetic rodents. Changes in Fgf21 expression due to suckling or nutritional manipulations were associated with changes in circulating free fatty acid and ketone body levels. In differentiated mouse brown adipocytes in culture, Fgf21 treatment increased the expression of thermogenic genes, caused higher total and uncoupled respiration, and enhanced glucose oxidation.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: FGF21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité