Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-FER Rabbit Polyclonal Antibody

Fournisseur: BosterBio
PA1882
BSBTPA1882EA 550 EUR InStock
BSBTPA1882
Anti-FER Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Tyrosine-protein kinase Fer(FER) detection. Tested with WB, IHC-P in Human;Rat.

FER(FPS/FES-Related tyrosine kinase) also known as TYK3, is an enzyme that in humans is encoded by the FER gene. Fer protein is a member of the FPS/FES family of nontransmembrane receptor tyrosine kinases. By in situ hybridization, Morris et al.(1990) concluded that the FER gene is located at 5q21-q22. Treatment of cells with JMP resulted in the release of FER from the cadherin complex and its accumulation in the integrin complex. The accumulation of FER in the integrin complex and the inhibitory effects of JMP could be reversed with a peptide that mimics the first coiled-coil domain of FER. The results suggested that FER mediates crosstalk between CDH2 and ITGB1. In Fer mutant mice, leukocyte emigration was exaggerated in response to LPS without altering vascular permeability, suggesting that FER has a role in regulating innate immunity.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

WB: The detection limit for FER is approximately 5ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: FER
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité