Anti-DDAH1 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB10000
BSBTPB10000EA 550 EUR InStock
BSBTPB10000
Anti-DDAH1 Rabbit Polyclonal Antibody
Anticorps
DDAH1 is knowns as dimethylarginine dimethylaminohydrolase 1 which is mapped to chromosome 1p22 by radiation hybrid and FISH analysis. This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. DDAH1 plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity. It widely expressed, especially in liver and kidney.
1. Dayoub, H., Achan, V., Adimoolam, S., Jacobi, J., Stuehlinger, M. C., Wang, B., Tsao, P. S., Kimoto, M., Vallance, P., Patterson, A. J., Cooke, J. P. Dimethylarginine dimethylaminohydrolase regulates nitric oxide synthesis: genetic and physiological evidence.Circulation 108: 3042-3047, 2003.
2. Millatt, L. J., Whitley, G. StJ., Li, D., Leiper, J. M., Siragy, H. M., Carey, R. M., Johns, R. A. Evidence for dysregulation of dimethylarginine dimethylaminohydrolase I in chronic hypoxia-induced pulmonary hypertension. Circulation 108: 1493-1498, 2003.
3. Tran, C. T. L., Fox, M. F., Vallance, P., Leiper, J. M. Chromosomal localization, gene structure, and expression pattern of DDAH1: comparison with DDAH2 and implications for evolutionary origins. Genomics 68: 101-105, 2000.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité