Anti-DDAH2 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB10001
BSBTPB10001EA 552 EUR InStock
BSBTPB10001
Anti-DDAH2 Rabbit Polyclonal Antibody
Anticorps
DDAH2 is known as dimethylarginine dimethylaminohydrolase 2 which is mapped to 6p21.3 by radiation hybrid and FISH analysis. This gene encodes a dimethylarginine dimethylaminohydrolase. DDAH2 functions in nitric oxide generation by regulating the cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity. The protein may be localized to the mitochondria. Alternative splicing resulting in multiple transcript variants.
1. Leiper, J. M,, Santa Maria, J., Chubb, A., MacAllister, R. J., Charles, I. G., Whitley, G. S., Vallance, P. Identification of two human dimethylarginine dimethylaminohydrolases with distinct tissue distributions and homology with microbial arginine deiminases.Biochem. J. 343: 209-214, 1999.
2. Smith, C. L., Birdsey, G. M., Anthony, S., Arrigoni, F. I., Leiper, J. M., Vallance, P. Dimethylarginine dimethylaminohydrolase activity modulates ADMA levels, VEGF expression, and cell phenotype. Biochem. Biophys. Res. Commun. 308: 984-989, 2003.
3. Tran, C. T. L., Fox, M. F., Vallance, P., Leiper, J. M. Chromosomal localization, gene structure, and expression pattern of DDAH1: comparison with DDAH2 and implications for evolutionary origins. Genomics 68: 101-105, 2000.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité