Anti-HMGB2 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB10002
BSBTPB10002EA 550 EUR InStock
BSBTPB10002
Anti-HMGB2 Rabbit Polyclonal Antibody
Anticorps
High-mobility group protein B2, also known as high-mobility group protein 2 (HMG-2), is a protein that in humans is encoded by the HMGB2 gene. This gene encodes a member of the non-histone chromosomal high mobility group protein family. The proteins of this family are chromatin-associated and ubiquitously distributed in the nucleus of higher eukaryotic cells. In vitro studies have demonstrated that this protein is able to efficiently bend DNA and form DNA circles. These studies suggest a role in facilitating cooperative interactions between cis-acting proteins by promoting DNA flexibility. This protein was also reported to be involved in the final ligation step in DNA end-joining processes of DNA double-strand breaks repair and V(D)J recombination.
1. "Entrez Gene: HMGB2 high-mobility group box 2".
2. Majumdar A, Brown D, Kerby S, Rudzinski I, Polte T, Randhawa Z, Seidman MM (Dec 1991). "Sequence of human HMG2 cDNA". Nucleic Acids Research 19 (23): 6643.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité