Anti-NAMPT Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB10009
BSBTPB10009EA 552 EUR InStock
BSBTPB10009
Anti-NAMPT Rabbit Polyclonal Antibody
Anticorps
Nicotinamide phosphoribosyltransferase (NAmPRTase or Nampt), also known as pre-B-cell colony-enhancing factor 1 (PBEF1) or visfatin, is an enzyme that in humans is encoded by the PBEF1 gene. This gene encodes a protein that catalyzes the condensation of nicotinamide with 5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, one step in the biosynthesis of nicotinamide adenine dinucleotide. The protein belongs to the nicotinic acid phosphoribosyltransferase (NAPRTase) family and is thought to be involved in many important biological processes, including metabolism, stress response and aging. This gene has a pseudogene on chromosome 10.
1. "Entrez Gene: PBEF1 pre-B-cell colony enhancing factor 1".
2. Samal B, Sun Y, Stearns G, Xie C, Suggs S, McNiece I (February 1994). "Cloning and characterization of the cDNA encoding a novel human pre-B-cell colony-enhancing factor". Mol. Cell. Biol. 14 (2): 1431–7.
3. Wang T, Zhang X, Bheda P, Revollo JR, Imai S, Wolberger C (July 2006). "Structure of Nampt/PBEF/visfatin, a mammalian NAD+ biosynthetic enzyme". Nat. Struct. Mol. Biol. 13 (7): 661–2.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité