Anti-VAMP7 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB10017
BSBTPB10017EA 579 EUR InStock
BSBTPB10017
Anti-VAMP7 Rabbit Polyclonal Antibody
Anticorps
Vesicle-associated membrane protein 7 (VAMP-7), also known as SYBL1, is a protein that in humans is encoded by the VAMP7 gene. This gene encodes a transmembrane protein that is a member of the soluble N-ethylmaleimide- sensitive factor attachment protein receptor (SNARE) family. The encoded protein localizes to late endosomes and lysosomes and is involved in the fusion of transport vesicles to their target membranes. Alternate splicing results in multiple transcript variants.
1. "Entrez Gene: SYBL1 synaptobrevin-like 1".
2. D'Esposito M, Ciccodicola A, Gianfrancesco F, Esposito T, Flagiello L, Mazzarella R, Schlessinger D, D'Urso M (Jul 1996). "A synaptobrevin-like gene in the Xq28 pseudoautosomal region undergoes X inactivation". Nat. Genet. 13 (2): 227–9.
3. Filippini F, Rossi V, Galli T, Budillon A, D'Urso M, D'Esposito M (Jul 2001). "Longins: a new evolutionary conserved VAMP family sharing a novel SNARE domain". Trends Biochem. Sci. 26 (7): 407–9.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité