Anti-BRCA1 Polyclonal Antibody
Fournisseur: Boster Bio
PB9015
BSBTPB9015EA 552 EUR InStock
BSBTPB9015
Anti-BRCA1 Polyclonal Antibody
Anticorps
Polyclonal antibody for BRCA1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. BRCA1 information: Molecular Weight: 207721 MW; Subcellular Localization: Nucleus . Chromosome . Cytoplasm . Localizes at sites of DNA damage at double-strand breaks (DSBs); recruitment to DNA damage sites is mediated by the BRCA1-A complex. Translocated to the cytoplasm during UV-induced apoptosis; Tissue Specificity: Isoform 1 and isoform 3 are widely expressed. Isoform 3 is reduced or absent in several breast and ovarian cancer cell lines.
Type: Primary
Antigen:More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité