Anti-CCL18/PARC Polyclonal Antibody
Fournisseur: Boster Bio
PB9023
BSBTPB9023EA 552 EUR InStock
BSBTPB9023
Anti-CCL18/PARC Polyclonal Antibody
Anticorps
Polyclonal antibody for PARC/CCL18 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Human. PARC/CCL18 information: Molecular Weight: 9849 MW; Subcellular Localization: Secreted; Tissue Specificity: Expressed at high levels in lung, lymph nodes, placenta, bone marrow, dendritic cells present in germinal centers and T-cell areas of secondary lymphoid organs and macrophages derived from peripheral blood monocytes. Not expressed by peripheral blood monocytes and a monocyte-to-macrophage differentiation is a prerequisite for expression. Expressed inMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité