Anti-Neuropilin-1 Polyclonal Antibody
Fournisseur: Boster Bio
PB9300
BSBTPB9300EA 552 EUR InStock
BSBTPB9300
Anti-Neuropilin-1 Polyclonal Antibody
Anticorps
Polyclonal antibody for NEUROPILIN 1/NRP1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. NEUROPILIN 1/NRP1 information: Molecular Weight: 103134 MW; Subcellular Localization: Cell membrane; Single-pass type I membrane protein; Tissue Specificity: The expression of isoforms 1 and 2 does not seem to overlap. Isoform 1 is expressed by the blood vessels of different tissues. In the developing embryo it is found predominantly in the nervous system. In adult tissues, it is highly expressed in heart and placenta; moderately in lung, liver, skeletalMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité