Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-AHR Rabbit Polyclonal Antibody

Fournisseur: BosterBio
PB9419
BSBTPB9419EA 579 EUR InStock
BSBTPB9419
Anti-AHR Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Aryl hydrocarbon receptor(AHR) detection. Tested with WB in Mouse;Rat.

AHR (aryl hydrocarbon receptor), also called bHLHe76, is a member of the family of basic helix-loop-helix transcription factors. AhR is a cytosolic transcription factor that is normally inactive, bound to several co-chaperones. The AHR gene is mapped on 7p21.1. Estrogenic actions of AHR agonists were detected in wildtype ovariectomized mouse uteri, but were absent in Ahr -/- or Er-alpha -/- ovariectomized mice. Complex assembly and ubiquitin ligase activity of CUL4B (AHR) in vitro and in vivo are dependent on the AHR ligand. In the CUL4B (AHR) complex, ligand-activated AHR acts as a substrate-specific adaptor component that targets sex steroid receptors for degradation. Cd4-positive cells from mice lacking Ahr developed Th17 responses but failed to produce Il22 and did not show enhanced Th17 development. Activation of Ahr during induction of EAE accelerated disease onset and increased pathology in wildtype mice, but not in Ahr -/- mice. The TDO-AHR pathway is active in human brain tumors and is associated with malignant progression and poor survival. Ahr activity within ROR-gamma-t-positive ILC could be induced by dietary ligands such as those contained in vegetables of the family Brassicaceae.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for AHR is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: ANXA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité