Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-YBX1 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
PB9465
BSBTPB9465EA 552 EUR InStock
BSBTPB9465
Anti-YBX1 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Nuclease-sensitive element-binding protein 1(YBX1) detection. Tested with WB, IHC-P, IHC-F in Human;Mouse;Rat.

YBX1(Y box binding protein 1), commonly referred to as "YB-1" by researchers, is a human protein. YB1 binding has an absolute requirement for the CCAAT box and relative specificity for the Y box. It has a molecular mass of 35,414 and contains 18% basic residues and putative nuclear localization signals. The YBX1 gene is mapped on 1p34.2. Ybx1 was highly expressed in mouse erythroid myeloid lymphoid clone-1 (EML), a hematopoietic precursor cell line, but that it was downregulated in myeloid progenitors and in Gmcsf-treated EML cells during myeloid differentiation. Ybx1 was expressed at high levels in myeloid leukemic cells at different developmental stages. Knockdown of YBX1 in a human leukemic cell line inhibited proliferation ability, induced apoptosis, and induced megakaryocytic differentiation in response to arsenic trioxide treatment. YBX1 is downregulated during myeloid differentiation and aberrant YBX1 expression in leukemic cells may contribute to leukemia development by blocking differentiation.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for YB1 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: YES1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité