Anti-HDAC7 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9629
BSBTPB9629EA 552 EUR InStock
BSBTPB9629
Anti-HDAC7 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Histone deacetylase 7(HDAC7) detection. Tested with WB in Human;Rat.
Histone deacetylase 7 is an enzyme that in humans is encoded by the HDAC7 gene. It is mapped to 12q13.1. Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation / deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene has sequence homology to members of the histone deacetylase family. This gene is orthologous to mouse HDAC7 gene whose protein promotes repression mediated via the transcriptional corepressor SMRT. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité