Anti-HSPA5 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9640
BSBTPB9640EA 552 EUR InStock
BSBTPB9640
Anti-HSPA5 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for 78 kDa glucose-regulated protein(HSPA5) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
HSPA5 (heat shock 70kDa protein 5), also known as glucose-regulated protein, 78kD (GRP78) or BiP, is a member of the heat-shock protein-70 (HSP70) family and is involved in the folding and assembly of proteins in the endoplasmic reticulum. BiP is also an essential component of the translocation machinery, as well as playing a role in retrograde transport across the ER membrane of aberrant proteins destined for degradation by the proteasome. The HSPA5 gene is mapped on 9q33.3. Shen et al. (2002) concluded that HSPA5 retains ATF6 in the ER by inhibiting its Golgi localization signals and that dissociation of HSPA5 during ER stress allows ATF6 to be transported to the Golgi. The findings of Shen et al. (2002) demonstrated that HSPA5 is a key element in sensing the folding capacity within the ER.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité