Anti-ING1 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9644
BSBTPB9644EA 552 EUR InStock
BSBTPB9644
Anti-ING1 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Inhibitor of growth protein 1(ING1) detection. Tested with WB in Human.
Inhibitor of growth protein 1 is a protein that in humans is encoded by the ING1 gene. It is mapped to 13q34. This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité