Anti-ITGA4 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9648
BSBTPB9648EA 552 EUR InStock
BSBTPB9648
Anti-ITGA4 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Integrin alpha-4(ITGA4) detection. Tested with WB, IHC-P in Human;Rat.
ITGA4/Integrin alpha4 is an integrin alpha subunit. It makes up half of the ?4?1 lymphocyte homing receptor. The product of this gene belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes an alpha 4 chain. Unlike other integrin alpha chains, alpha 4 neither contains an I-domain, nor undergoes disulfide-linked cleavage. Alpha 4 chain associates with either beta 1 chain or beta 7 chain. ITGA4 is also mapped to 2q31-q32 by fluorescence in situ hybridization.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité