Anti-LCAT Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9657
BSBTPB9657EA 552 EUR InStock
BSBTPB9657
Anti-LCAT Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Phosphatidylcholine-sterol acyltransferase(LCAT) detection. Tested with WB in Human;Mouse;Rat.
LCAT (Lecithin: Cholesterol Acyltransferase), is an enzyme that converts free cholesterol into cholesteryl ester. Azoulay et al. (1987) used a cDNA clone corresponding to LCAT to assign the locus to 16q22 through the analysis of DNA from somatic cell hybrids and in situ hybridization. LCAT plays an important role in lipoprotein metabolism, especially in the process termed 'reverse cholesterol transport.' The enzyme is synthesized in the liver and circulates in blood plasma as a complex with components of high density lipoprotein (HDL). Cholesterol from peripheral cells is transferred to HDL particles, esterified through the action of LCAT on HDL, and incorporated into the core of the lipoprotein. The cholesterol ester is thereby transported to the liver (Jonas, 2000).
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité