Anti-LGALS8 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9659
BSBTPB9659EA 552 EUR InStock
BSBTPB9659
Anti-LGALS8 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Galectin-8(LGALS8) detection. Tested with WB in Human;Rat.
Galectin-8 is a protein of the galectin family that in humans is encoded by the LGALS8 gene. This gene encodes a member of the galectin family. Galectins are beta-galactoside-binding animal lectins with conserved carbohydrate recognition domains. The galectins have been implicated in many essential functions including development, differentiation, cell-cell adhesion, cell-matrix interaction, growth regulation, apoptosis, and RNA splicing. This gene is widely expressed in tumoral tissues and seems to be involved in integrin-like cell interactions. Alternatively spliced transcript variants encoding different isoforms have been identified.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité