Anti-MAPK6 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9666
BSBTPB9666EA 552 EUR InStock
BSBTPB9666
Anti-MAPK6 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Mitogen-activated protein kinase 6(MAPK6) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Mitogen-activated protein kinase 6 is an enzyme that in humans is encoded by the MAPK6 gene. It is mapped to 15q21. The protein encoded by this gene is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases). MAP kinases also known as extracellular signal-regulated kinases (ERKs), are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals. This kinase is localized in the nucleus, and has been reported to be activated in fibroblastsupon treatment with serum or phorbol esters.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité