Anti-NDRG2 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9672
BSBTPB9672EA 552 EUR InStock
BSBTPB9672
Anti-NDRG2 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Protein NDRG2(NDRG2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Protein NDRG2, also known as KIAA1248, is a protein that in humans is encoded by the NDRG2 gene. It is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. This gene is mapped to 14q11.2. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. It contributes to the regulation of the WNT signaling pathway. Also, this gene may be involved in dendritic cell and neuron differentiation.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité