Anti-SECTM1 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9679
BSBTPB9679EA 579 EUR InStock
BSBTPB9679
Anti-SECTM1 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Secreted and transmembrane protein 1b(SECTM1) detection. Tested with WB in Mouse.
SECTM1B is also known as Sectm1 or K12. Secreted and transmembrane protein 1 is a protein that in humans is encoded by the SECTM1 gene. This gene encodes a transmembrane and secreted protein with characteristics of a type 1a transmembrane protein. It is found in a perinuclear Golgi-like pattern and thought to be involved in hematopoietic and/or immune system processes.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité