Anti-AEBP2 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9977
BSBTPB9977EA 552 EUR InStock
BSBTPB9977
Anti-AEBP2 Rabbit Polyclonal Antibody
Anticorps
Adipocyte Enhancer-Binding Protein is a zinc finger protein that in humans is encoded by the evolutionarily well-conserved gene AEBP2. This gene is mapped to 12p12.3. AEBP2 is a DNA-binding transcriptional repressor. It may regulate the migration and development of the neural crest cells through the PRC2-mediated epigenetic mechanism and is most likely a targeting protein for the mammalian PRC2 complex.
1. Imhof, Axel; Kim, Hana; Bakshi, Arundhati; Kim, Joomyeong (2015). "Retrotransposon-Derived Promoter of Mammalian Aebp2". PLOS ONE 10 (4): e0126966.
2. Kim H, Kang K, Kim J (2009). "AEBP2 as a potential targeting protein for Polycomb Repression Complex PRC2". Nucleic Acids Res. 37(9): 2940–50.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité