Anti-AGO4 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9979
BSBTPB9979EA 579 EUR InStock
BSBTPB9979
Anti-AGO4 Rabbit Polyclonal Antibody
Anticorps
AGO4 (Argonaute 4) is also known as EIF2C4. This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic containing PAZ and PIWI domains, and it may play a role in short-interfering-RNA-mediated gene silencing. This gene is located on chromosome 1 in a cluster of closely related family members including argonaute 3, and eukaryotic translation initiation factor 2C, 1.
1. Carmell, M. A., Xuan, Z., Zhang, M. Q., Hannon, G. J. The argonaute family: tentacles that reach into RNAi, developmental control, stem cell maintenance, and tumorigenesis. Genes Dev. 16: 2733-2742, 2002.
2. Nagase, T., Kikuno, R., Nakayama, M., Hirosawa, M., Ohara, O. Prediction of the coding sequences of unidentified human genes. XVIII. The complete sequences of 100 new cDNA clones from brain which code for large proteins in vitro. DNA Res. 7: 273-281, 2000.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité