Anti-AMACR Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9983
BSBTPB9983EA 552 EUR InStock
BSBTPB9983
Anti-AMACR Rabbit Polyclonal Antibody
Anticorps
Alpha-methylacyl-CoA racemase (AMACR) is a mitochondrial and peroxisomal enzyme. It encodes a racemase. The encoded enzyme interconverts pristanoyl-CoA and C27-bile acylCoAs between their (R)- and (S)-stereoisomers. The conversion to the (S)-stereoisomers is necessary for degradation of these substrates by peroxisomal beta-oxidation. Encoded proteins from this locus localize to both mitochondria and peroxisomes. Mutations in this gene may be associated with adult-onset sensorimotor neuropathy, pigmentary retinopathy, and adrenomyeloneuropathy due to defects in bile acid synthesis. Alternatively spliced transcript variants have been described. Read-through transcription also exists between this gene and the upstream neighboring C1QTNF3 (C1q and tumor necrosis factor related protein 3) gene.
1. Amery, L., Fransen, M., De Nys, K., Mannaerts, G. P., Van Veldhoven, P. P. Mitochondrial and peroxisomal targeting of 2-methylacyl-CoA racemase in humans. J. Lipid Res. 41: 1752-1759, 2000.
2. Clarke, C. E., Alger, S., Preece, M. A., Burdon, M. A., Chavda, S., Denis, S., Ferdinandusse, S., Wanders, R. J. A. Tremor and deep white matter changes in alpha-methylacyl-CoA racemase deficiency. Neurology 63: 188-189, 2004.
3. Dick, D., Horvath, R., Chinnery, P. F. AMACR mutations cause late-onset autosomal recessive cerebellar ataxia. Neurology 76: 1768-1770, 2011.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité