Anti-AMHR2 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9984
BSBTPB9984EA 550 EUR InStock
BSBTPB9984
Anti-AMHR2 Rabbit Polyclonal Antibody
Anticorps
AMHR2 is the receptor for the anti-Mullerian hormone (AMH) which, in addition to testosterone, results in male sex differentiation. AMH and testosterone are produced in the testes by different cells and have different effects. Testosterone promotes the development of male genitalia while the binding of AMH to the encoded receptor prevents the development of the mullerian ducts into uterus and Fallopian tubes. Mutations in this gene are associated with persistent Mullerian duct syndrome type II. Alternatively spliced transcript variants encoding different isoforms have been identified.
1. Behringer RR (1994). "The in vivo roles of müllerian-inhibiting substance". Current Topics in Developmental Biology. Current Topics in Developmental Biology 29: 171–87.
2. Cate RL, Mattaliano RJ, Hession C, Tizard R, Farber NM, Cheung A, Ninfa EG, Frey AZ, Gash DJ, Chow EP (Jun 1986). "Isolation of the bovine and human genes for Müllerian inhibiting substance and expression of the human gene in animal cells". Cell 45 (5): 685–98.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité