Anti-APOE Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9986
BSBTPB9986EA 552 EUR InStock
BSBTPB9986
Anti-APOE Rabbit Polyclonal Antibody
Anticorps
APOE is also known as AD2 or LPG. The protein encoded by this gene is a major apoprotein of the chylomicron. It binds to a specific liver and peripheral cell receptor, and is essential for the normal catabolism of triglyceride-rich lipoprotein constituents. This gene maps to chromosome 19 in a cluster with the related apolipoprotein C1 and C2 genes. Mutations in this gene result in familial dysbetalipoproteinemia, or type III hyperlipoproteinemia (HLP III), in which increased plasma cholesterol and triglycerides are the consequence of impaired clearance of chylomicron and VLDL remnants. Alternative splicing results in multiple transcript variants.
1. APOE ? variants increase risk of warfarin-related intracerebral hemorrhage.Falcone GJ, et al. Neurology, 2014 Sep 23.
2. Calabuig-Navarro MV, et al. Apolipoprotein E genotype has a modest impact on the postprandial plasma response to meals of varying fat composition in healthy men in a randomized controlled trial. J Nutr, 2014 Nov.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité