Anti-CCT2 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9992
BSBTPB9992EA 552 EUR InStock
BSBTPB9992
Anti-CCT2 Rabbit Polyclonal Antibody
Anticorps
T-complex protein 1 subunit beta (TCP1 beta) is a protein that in humans is encoded by the CCT2 gene. The protein encoded by this gene is a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Two transcript variants encoding different isoforms have been found for this gene.
1. "Entrez Gene: CCT2 chaperonin containing TCP1, subunit 2 (beta)".
2. Won KA, Schumacher RJ, Farr GW, Horwich AL, Reed SI (Dec 1998). "Maturation of human cyclin E requires the function of eukaryotic chaperonin CCT". Molecular and Cellular Biology 18 (12): 7584–9.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité