Anti-CCT8 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9993
BSBTPB9993EA 552 EUR InStock
BSBTPB9993
Anti-CCT8 Rabbit Polyclonal Antibody
Anticorps
T-complex protein 1 subunit theta is a protein that in humans is encoded by the CCT8 gene. This gene encodes the theta subunit of the CCT chaperonin, which is abundant in the eukaryotic cytosol and may be involved in the transport and assembly of newly synthesized proteins. Alternative splicing results in multiple transcript variants of this gene. A pseudogene related to this gene is located on chromosome 1.
1. "Entrez Gene: CCT8 chaperonin containing TCP1, subunit 8 (theta)".
2. Kubota H, Hynes G, Willison K (Apr 1995). "The eighth Cct gene, Cctq, encoding the theta subunit of the cytosolic chaperonin containing TCP-1". Gene 154 (2): 231–6.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité