Anti-CD274 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9994
BSBTPB9994EA 579 EUR InStock
BSBTPB9994
Anti-CD274 Rabbit Polyclonal Antibody
Anticorps
Programmed death-ligand 1 (PD-L1), also known as CD274 or B7-H1, is a protein that in humans is encoded by the CD274 gene. It is mapped to 9p24.1. PD-L1 is a 40kDa type 1 transmembrane protein that has been speculated to play a major role in suppressing the immune system during particular events such as pregnancy, tissue allografts, autoimmune disease and other disease states such as hepatitis. It has been concluded that upregulation of PD-L1 on tumor MDCs downregulates T-cell immunity and that PD-L1 blockade may represent an approach for cancer immunotherapy. Additionally, PD-L1 can provide positive costimulatory signals for innate and adaptive immunity and for protection against intracellular bacterial infection. What’s more, it has been found that PD1/PDL1 pathway may be a good target for restoring antitumor immunity in ovarian cancer.
1. Barber, D. L., Wherry, E. J., Masopust, D., Zhu, B., Allison, J. P., Sharpe, A. H., Freeman, G. J., Ahmed, R. Restoring function in exhausted CD8 T cells during chronic viral infection. Nature 439: 682-687, 2006.
2. Dong, H., Zhu, G., Tamada, K., Chen, L. B7-H1, a third member of the B7 family, co-stimulates T-cell proliferation and interleukin-10 secretion.Nature Med. 5: 1365-1369, 1999.
3. Curiel, T. J., Wei, S., Dong, H., Alvarez, X., Cheng, P., Mottram, P., Krzysiek, R., Knutson, K. L., Daniel, B., Zimmermann, M. C., David, O., Burow, M., and 10 others. Blockade of B7-H1 improves myeloid dendritic cell-mediated antitumor immunity. Nature Med. 9: 562-567, 2003.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité