Anti-CDK6 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9995
BSBTPB9995EA 552 EUR InStock
BSBTPB9995
Anti-CDK6 Rabbit Polyclonal Antibody
Anticorps
Cell division protein kinase 6, also called Plstire, is an enzyme that in humans is encoded by the CDK6 gene. The protein encoded by this gene is a member of the cyclin-dependent protein kinase (CDK) family. Radiation hybrid analysis and inclusion within a mapped clone place the CDK6 gene at 7q21. Serine/threonine-protein kinase involved in the control of the cell cycle and differentiation and promotes G1/S transition. This gene also involved in initiation and maintenance of cell cycle exit during cell differentiation. It prevents cell proliferation and regulates negatively cell differentiation, but is required for the proliferation of specific cell types. In addition, CDK6 plays a role in promoting the proliferation of beta-cells in pancreatic islets of Langerhans.
1. Lena, A. M., Mancini, M., Rivetti di Val Cervo, P. R., Saintigny, G., Mahe, C., Melino, G., Candi, E. MicroRNA-191 triggers keratinocytes senescence by SATB1 and CDK6 downregulation. Biochem. Biophys. Res. Commun. 423: 509-514, 2012.
2. Lien, H.-C., Lin, C.-W., Huang, P.-H., Chang, M.-L., Hsu, S.-M. Expression of cyclin-dependent kinase 6 (cdk6) and frequent loss of CD44 in nasal-nasopharyngeal NK/T-cell lymphomas: comparison with CD56-negative peripheral T-cell lymphomas. Lab. Invest. 80: 893-900, 2000.
3. Malumbres, M., Sotillo, R., Santamaria, D., Galan, J., Cerezo, A., Ortega, S., Dubus, P., Barbacid, M. Mammalian cells cycle without the D-type cyclin-dependent kinases Cdk4 and Cdk6. Cell 118: 493-504, 2004.
4. Veiga-Fernandes, H., Rocha, B. High expression of active CDK6 in the cytoplasm of CD8 memory cells favors rapid division. Nature Immun. 5: 31-37, 2003.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité