proBDNF Rapid ELISA Kit: Human, Mouse, Rat (2 Plates)
Fournisseur: Biosensis
BEK-2237-2P
BSENBEK-2237-2PEA 731 EUR InStock
BSENBEK-2237-2P
proBDNF Rapid ELISA Kit: Human, Mouse, Rat (2 Plates)
Tests Tests ELISA
Biosensis proBDNF (Brain-derived neurotrophic factor, BDNF, Abrineurin) RapidTM enzyme-linked immunosorbent assay (ELISA) Kit is a sandwich ELISA that allows the specific, fast and reliable quantification of proBDNF in less than 4 hours in cell culture supernatants, human serum and EDTA-plasma only if used as directed. Please refer to the kit protocol for specific use instructions for each substrate application, in particular human blood samples.
This ELISA kit consists of a pre-coated monoclonal anti-proBDNF capture antibody, a biotinylated anti-matureBDNF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of proBDNF present in samples and protein standards. A proBDNF positive control (QC sample) is provided to assure consistent assay performance.
This proBDNF ELISA kit employs a recombinant, cleavage-resistant human proBDNF standard produced in mammalian cells by Biosensis and validated against externally available proBDNF proteins.
Note that accurate proBDNF quantification in human serum and EDTA-plasma requires the addition of Heterophilic Antibody Blocker BL-004-500 provided in the kit, and available for purchase separately.
This ELISA kit has not been tested for other applications. It has been configured for research use only and is not to be used for diagnostic or clinical procedures. The ELISA kit box contains 2× 96 pre-coated strip plate, protein standards, QC sample, detection reagents, heterophilic antibody blocker, wash and sample buffers and detailed protocols.
Range: 15,6 to 1000 pg/ml; Typical limit of detection (LOD) for proBDNF is 6 pg/ml determined as 150% of the blank value.
Human proBDNF. The capture antibody used in this ELISA kit binds to an epitope within the pro-domain of proBDNF. Thus, this ELISA detects the full length form of proBDNF and does not quantify mature BDNF. Whether this ELISA kit can detect truncated BDNF is unknown at present. Mature BDNF cross-reactivity was assessed by spiking 28 kDa mature BDNF protein obtained from WHO (www.nibsc.org) into a 1/5 diluted human serum sample at 5 ng/ml, which represents a BDNF concentration level of 25 ng/ml in undiluted, normal human serum. Cross-reactivity of mature BDNF was < 0,3% (w/v), or < 0,1% in molar concentration. The assay antibodies do not cross-react with nerve growth factor (NGF), neurotrophin-3 (NT-3) or NT-4/5. Species cross-reactivity has not yet been determined.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité