Anti-Peripherin Chicken Polyclonal Antibody
Fournisseur: Biosensis
C-1399-50
BSENC-1399-50EA 312 EUR InStock
BSENC-1399-50
Anti-Peripherin Chicken Polyclonal Antibody
Anticorps
Peripherin is a class-III neuronal intermediate filament protein found in certain classes of neuron, most of which are located in the peripheral nervous system. Peripherin is a ~57kDa intermediate filament subunit found initially in sensory neurons of the peripheral nervous systems, which gives the protein its name. The HGNC name for this protein is PRPH. Subsequently, peripherin was found in some sensory and other neurons of the central nervous system and also in rat pheochromacytoma PC12 cells. Peripherin is also expressed in certain neuroendocrine tumors and in the insulin producing cells of the pancreas. Peripherin belongs to the Class III family of intermediate filament subunits which also includes vimentin, glial fibrillary acidic protein (GFAP) and desmin.
The specificity of this antibody has been confirmed by WB. This antibody detects ~57 kDa Peripherin protein in rat brain stem and spinal cord extracts. A suitable control tissue is rat spinal cord or peripheral nerve homogenate.
Application Information:
Western Blotting (WB) and Immunocytochemistry (IC). A dilution of 1:5,000 - 1:10,000 is recommended for WB. A dilution of 1:500-1,000 is recommended for IC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Peripherin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Human, Rat, Mouse, Cat, Mammals
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité