Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-Ubiquitin C Terminal Hydrolase 1 Chicken Polyclonal Antibody

Fournisseur: Biosensis
C-1406-50
BSENC-1406-50EA 312 EUR InStock
BSENC-1406-50
Anti-Ubiquitin C Terminal Hydrolase 1 Chicken Polyclonal Antibody
Anticorps
Ubiquitin C-terminal hydrolase 1 (UCHL1) has several other names, such as ubiquitin carboxyl esterase L1, ubiquitin thiolesterase, neuron-specific protein PGP9.5 and Park5. It was originally identified as a major component of the neuronal cytoplasm from 2-dimensional gel analysis of brain tissues, and was given the name PGP9.5 (1). The protein is extremely abundant, and was estimated to be present at a concentration of 200-500 µg/g wet weight, representing a major protein component of neuronal cytoplasm (1). This has been claimed to represent 1-2% of total brain protein. It was later found that a ubiquitin C-terminal hydrolase enzyme activity was associated with the PGP9.5 protein, resulting in the renaming of PGP9.5 to ubiquitin C-terminal hydrolase 1.This enzyme is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. The enzyme also binds to free monoubiquitin and may prevent its degradation in lysosomes.

The specificity of this antibody has been confirmed by WB. This antibody detects ~24 kDa UCHL1 enzyme. Suitable control tissue is rat spinal cord or peripheral nerve homogenate. In cell staining, the antibody stains neurons strongly and specifically, and the staining is concentrated in the cell bodies, though some does extend into the dendrites.
Application Information:
Western Blotting (WB) and Immunocytochemistry (IC). A dilution of 1:5,000 - 1:10,000 is recommended for WB. A dilution of 1:500-1,000 is recommended for IC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Ubiquitin C Terminal Hydrolase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Human, Rat, Mouse, Bovine, Pig

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité