Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

L'expérience en ligne d'Avantor évolue pour mieux vous accompagner ! À compter du 4 avril 2025, nos clients seront migrés vers une nouvelle plateforme pour une expérience d'achat en ligne simplifiée.

Neurotrophin antibody kit

Fournisseur: Biosensis

KIT-NT-100 KIT-NT-500
BSENKIT-NT-100EA 302 EUR InStock
BSENKIT-NT-100 BSENKIT-NT-500
Neurotrophin antibody kit
Tests Tests immunologiques
The Neurotrophin Antibody Kit contains a collection of sheep antibodies raised against BDNF, NGF, NT3 and NT4/5. The kit is completed with one vial of normal sheep IgG for use as negative control. This kit presents a cost-effective way to investigate neurotrophin protein expression and function by immunological techniques such as immunohistochemistry, ELISA, and inhibition of biological activity.

Neurotrophins are growth factors that regulate neuronal development, maintenance and plasticity. These homo-dimeric proteins activate a multitude of intracellular signalling cascades through their interaction with Trk receptors and p75NTR.

Applications:
Immunohistochemistry: 1-10 µg/ml. BDNF antibody works best on Zamboni's fixed, frozen tissue, and is not recommended for paraffin-embedded tissue.
ELISA: 1-10 µg/ml.
Western Blotting: 1-10 µg/ml. Western blotting is not recommended for the BDNF antibody.
Inhibition of biological activity in vitro/in vivo: Expected ED50 (in vivo) values are in the range of 2-10 µg/ml. The NGF antibody completely inhibits neuronal survival and the outgrowth actions of murine NGF in chicken DRG in vitro.

Biosensis recommends optimal dilutions/concentrations should be determined by the end user. The negative control (normal sheep IgG) should be used at the same concentration as the neurotrophin antibodies.

Specificity:
BDNF antibody: Cross-reactivity of <1% against mouse NGF, recombinant human NT3 or NT4/5 has been shown by ELISA.
NGF antibody: Cross-reactivity of <1% against recombinant human BDNF, NT3 and NT4/5 by ELISA has been shown
NT3 antibody: Cross-reactivity of <1% to mouse NGF, recombinant human BDNF and <5% to NT4/5 has been shown by ELISA.
NT4/5 antibody: <1% cross-reactivity against NGF, recombinant human BDNF and <5% to NT3 has been shown by dot blot.

Reconstitute each vial with sterile-filtered, ultrapure water, to achieve an antibody concentration of 1 mg/ml and centrifuge briefly to remove any insoluble material.

Information de livraison: Neurotrophin Antibody Kit (trial size) contains 100 μg sheep IgG of each antibody. Full size kit contains 500 μg sheep IgG of each antibody.

Attention: Note: Store lyophilised, unopened vials at 2 to 8 °C or lower. After reconstitution, prepare aliquots and store at –20 °C for a higher stability. Avoid freeze-thaw cycles.


Commander maintenant

 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité

Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.

Pour en savoir plus, consultez notre Politique de Confidentialit&eacute;..