Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-p75NTR Mouse Monoclonal Antibody [clone: ME20.4]

Fournisseur: Biosensis
M-011-100
BSENM-011-100EA 312 EUR InStock
BSENM-011-100
Anti-p75NTR Mouse Monoclonal Antibody [clone: ME20.4]
Anticorps
Nerve growth factor receptor (NGFR) is also referred to as p75(NTR) due to its molecular mass and its ability to bind at low affinity not only NGF (see 162030), but also other neurotrophins, including brain-derived neurotrophic factor (BDNF; 113505), neurotrophin-3 (NTF3; 162660), and neurotrophin-4/5 (NTF5; 162662). At the time of its discovery, NGFR was considered a unique type of protein. Subsequently, however, a large superfamily of tumor necrosis factor receptors were found to share the overall structure of NGFR (4 extracellular ligand-binding, cysteine-rich repeats, or CRs, and signaling through association with, or disassociation from, cytoplasmic interactors). The identification of this superfamily helped elucidate some of the biologic functions of NGFR, including its ultimate involvement in the nuclear factor kappa-B (NFKB; see 164011) and apoptosis pathways. As a monomer, NGFR binds NGF with low affinity. Higher affinity binding is achieved by association with higher molecular mass, low-affinity neurotrophin receptors, namely the tropomyosin receptor kinases, TRKA (NTRK1; 191315), TRKB (NTRK2; 600456), and TRKC (NTRK3; 191316). TRKA, TRKB, and TRKC are specific for or 'preferred by' NGF, NTF5 and BDNF, and NTF3, respectively (Ip et al., 1993). NTF3 also binds to TRKA and TRKB, but with significantly lower affinity

This antibody recognises p75NTR (low affinity neurotrophin receptor)
Application Information:
Immunohistochemistry, immunofluorescence, flow cytometry. Suggested working dilutions: For Immunohistochemistry a concentration of 2 µg/ml is recommended. Antibody not appropriate for Western Blot. For FACS a concentration of 20 µg/ml is recommended. At least 1 in 5000 dilution is recommended for 1 site ELISAs. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: p75NTR
Clonality: Monoclonal
Clone: ME20.4
Conjugation: Unconjugated
Epitope: Human NT3 (19-128)
Host: Mouse
Isotype:
Reactivity: Human, Cat, Dog, Pig, Rabbit, Sheep

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité