Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-TNR16 Mouse Monoclonal Antibody [clone: Clone: Me20.4] (Atto 488)

Fournisseur: Biosensis
M-027-50-AT
BSENM-027-50-ATEA 302 EUR InStock
BSENM-027-50-AT
Anti-TNR16 Mouse Monoclonal Antibody [clone: Clone: Me20.4] (Atto 488)
Anticorps
Nerve growth factor receptor (NGFR) is also referred to as p75(NTR) due to its molecular mass and its ability to bind at low affinity not only NGF (see 162030), but also other neurotrophins, including brain-derived neurotrophic factor (BDNF; 113505), neurotrophin-3 (NTF3; 162660), and neurotrophin-4/5 (NTF5; 162662). At the time of its discovery, NGFR was considered a unique type of protein. Subsequently, however, a large superfamily of tumor necrosis factor receptors were found to share the overall structure of NGFR (4 extracellular ligand-binding, cysteine-rich repeats, or CRs, and signaling through association with, or disassociation from, cytoplasmic interactors). The identification of this superfamily helped elucidate some of the biologic functions of NGFR, including its ultimate involvement in the nuclear factor kappa-B (NFKB; see 164011) and apoptosis pathways. As a monomer, NGFR binds NGF with low affinity. Higher affinity binding is achieved by association with higher molecular mass, low-affinity neurotrophin receptors, namely the tropomyosin receptor kinases, TRKA (NTRK1; 191315), TRKB (NTRK2; 600456), and TRKC (NTRK3; 191316). TRKA, TRKB, and TRKC are specific for or 'preferred by' NGF, NTF5 and BDNF, and NTF3, respectively (Ip et al., 1993). NTF3 also binds to TRKA and TRKB, but with significantly lower affinity.
Application information:
This antibody is recommended for use in immunohistochemistry, immunofluorescence, flow cytometry and NGF receptor p75 dynamics. For immunohistochemistry a concentration of 2 µg/mL is recommended. Not appropriate for Western Blots. For FACS a concentration of 20 µg/mL is recommended and for 1 site ELISA at least a 1 in 5000 dilution. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNR16
Clonality: Monoclonal
Clone: Clone: Me20.4
Conjugation: Atto 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, cat, dog, pig, rabbit, sheep

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité