Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-Ubiquitin Mouse Monoclonal Antibody [clone: Ubi-1]

Fournisseur: Biosensis
M-1404-100
BSENM-1404-100EA 312 EUR InStock
BSENM-1404-100
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: Ubi-1]
Anticorps
Ubiquitin is a highly conserved 76 amino acid protein with an estimated molecular weight of 8.56 kDa which has a central role in regulated protein degradation. It is a protein modifier which can be covalently attached to target lysines either as a monomer or as a lysine-linked polymer. Several types of polymeric chains can be formed depending on the lysine used for the assembly. Attachment to proteins as a polymer leads to their degradation by the 26S proteosome; a complex, multicatalytic cytosolic and nuclear protease. Attachment to proteins as a monomer or as an alternatively linked polymer does not lead to proteasomal degradation and may be required for numerous functions, including maintenance of chromatic structure, regulation of gene expression, stress response, ribosome biogenesis and DNA repair. Ubiquitin is synthesized as a polyubiquitin precursor with exact head to tail repeats, the number of repeats of which differ between species and strains. In some species there is a final amino-acid after the last repeat, here in bovine a Cys. Some ubiquitin genes contain a single copy of ubiquitin fused to a ribosomal protein (either L40 or S27a).

The specificity of this antibody has been confirmed by WB. This antibody detects ~8.5 kDa Ubiquitin.
Application Information:
Western Blotting (WB), Immunohistochemistry - paraffin embedded tissue (IH-P) and ELISA. Suggested dilution for WB is 1:500-1,000. This antibody can be used on mildly fixed histological sections of human brain for studies of Alzheimer's disease. This antibody also works on paraffin embedded material. It also recognises other ubiquinated inclusion bodies such as Lewy bodies of Parkinson's disease and the Pick bodies in Pick's disease in formalin fixed tissues. Suggested dilution for IH is 1:500. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Ubiquitin
Clonality: Monoclonal
Clone: Ubi-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Chicken, Drosophila, C. elegans

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité