Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: LNC 1; LNC1;LNC-1]

Fournisseur: Biosensis
M-1616-100
BSENM-1616-100EA 312 EUR InStock
BSENM-1616-100
Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: LNC 1; LNC1;LNC-1]
Anticorps
Tyrosine hydroxylase is an excellent marker for dopaminergic and noradrenergic neurons. Tyrosine hydroxylase (a.k.a. tyrosine 3-monooxygenase) is the enzyme responsible for catalyzing the conversion of the amino acid L-tyrosine to L-3,4-dihydroxyphenylalanine (L-DOPA). L-DOPA is a precursor for dopamine, which, in turn, is a precursor for the important neurotransmitters norepinephrine (noradrenaline) and epinephrine (adrenaline). Tyrosine hydroxylase catalyzes the rate limiting step in this synthesis of catecholamines. In humans, tyrosine hydroxylase is encoded by the TH gene, and the enzyme is present in the central nervous system (CNS), peripheral symphatic neurons and the adrenal medulla. The enzymatic activity of TH requires ferrous ions as cofactors and is believed to be regulated by phosphorylation. At least four isoforms of human TH have been identified which result from alternative splicing. Tyrosine hydroxylase, phenylalanine hydroxylase and tryptophan hydroxylase together make up the family of aromatic amino acid hydroxylases (AAAHs). http://en.wikipedia.org/wiki/Tyrosine_hydroxylase

Clone LNC 1 recognizes an epitope on the outside of the regulatory N-terminus. The clone detects a protein of approximately 59-61 kDa by Western blot and reduced SDS-PAGE. The clone does not react with dopamine-beta-hydroxylase, phenylalanine hydroxylase, trytophan hydroxylase, dehydropteridine reductase, sepiapterin reductase or phenethanolamine-N-methyl transferase (PNMT) by western blots.
Application Information:
Western Blotting (WB), Immunohistochemistry (IH), Immunohistochemistry/paraffin embedded IH(P), Immunoprecipitation (IP), Immunofluorescence (IF), Flow cytometry (FC).WB: 1:1000 -1:2000, SDS reduced samples. Detects a 59-61kDa protein. Rat Brain lysates is a suitable control.IHC/IH(P): Reacts in formalin fixed paraffin embedded tissues with HIER antigen recovery. Typical dilution is 1:100-1:200 depending upon incubation time and detection method used. IF: 1:200-1:1000, 4% PFA fixed tissues/cells permeabilized with 0.1-0.4% triton X-100; also works in fresh frozen and acetone fixed tissues/cells.IP: 1:100, immobilized on protein A beads, Fleming-Jones et al (1995) J. Protein Chemistry 14(5):275-282.FC: Fixed, permeabilized dopaminergic nerve terminals from rat striatum, {Wolf, ME, Kapatos, G (1989) The Journal of Neuroscience, January 1989, 9(l): 108-114; Wolf ME, Zigmond, MJ, Kapatos, G (1989) J. Neurochemistry 53(3):879-885}.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Monoclonal
Clone: LNC 1; LNC1;LNC-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken, Frog, Horse, Human, Monkey, Mouse, Vole, Sheep, Zebrafish

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité