Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-VISL1 Mouse T4 Antibody [clone: 2D11]

Fournisseur: Biosensis
M-1657-100
BSENM-1657-100EA 312 EUR InStock
BSENM-1657-100
Anti-VISL1 Mouse T4 Antibody [clone: 2D11]
Anticorps
Visinin (sometimes known as hippocalcin-like protein 3, HLP3, HPCAL3, HUVISL1, VLP-1, VILIP and VILIP-1) was originally isolated biochemically from chicken retina as a major protein of about 24kDa on SDS-PAGE. Following cloning and sequencing of visinin, several visinin like proteins were discovered by homology screening. One of these, Visinin-like protein 1 is a small Calcium binding protein which is very abundant in the nervous system and is found only in neurons, though different neurons have different levels of expression. It is particularly concentrated in cerebellar Purkinje cells, and tends to be most abundant in perikarya and dendrites. The protein belongs to the large superfamly of calmodulin and paravalbumin type proteins which function by binding Calcium ions. Calcium binding alters the confomation of these proteins and allow them to interact with other binding partners, the properties of which they may alter. Visinin-like protein 1 has four 'EF hand' domains, which are negatively charged helix-turn-helix peptides which are responsible for Calcium binding. Visinin-like protein 1 is 191 amino acids in size and has a molecular weight on SDS-PAGE of 22kDa.

  • The antibody reacts with a 22 kDa band by Western blot on bovine cerebellum lysate

Application information: Western Blotting (WB), Immunocytochemistry (ICC) and Immunohistochemistry (IHC). A dilution of 1:500 to 1:1,000 is recommended for WB. A dilution of 1:500 to 1:1,000 is recommended for IHC and ICC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: VISL1
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat, Mouse, Cow

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité