Anti-NTRK2 Mouse Monoclonal Antibody [clone: BS379]
Fournisseur: Biosensis
M-1836-100
BSENM-1836-100EA 302 EUR InStock
BSENM-1836-100
Anti-NTRK2 Mouse Monoclonal Antibody [clone: BS379]
Anticorps
The protein named TrkB (also named Neurotrophic tyrosine kinase receptor type 2 (NTRK2), GP145-TrkB or Tropomyosin-related kinase B is a receptor tyrosine kinase involved in the development and the maturation of the central and the peripheral nervous systems and is important in the regulation of neuron survival, proliferation, migration, differentiation, and synapse formation and plasticity. TrkB may also play a role in neutrophin-dependent calcium signaling in glial cells and mediate communication between neurons and glia. TrkB is the primary receptor for BDNF (brain-derived neurotrophic factor. TrkB also binds NT4 and NT3 but less efficiently. (Reference: www.uniprot.org).
Applications: Flow Cytometry (5-10 ug/mL): Tested on human and rodent cell lines. Cell staining can be performed under native conditions on ice, or on fixed cells with up to 4% formaldehyde. Other applications have not been tested. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application notes: Flow Cytometry (5-10 ug/mL): Tested on human and rodent cell lines. Cell staining can be performed under native conditions on ice, or on fixed cells with up to 4% formaldehyde.
Other applications have not been tested. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité