Saponaria officinalis Native Saporin (from Saponaria officinalis seeds)
Fournisseur: Biosensis
PE-001-100
BSENPE-001-100EA 140 EUR InStock
BSENPE-001-100
Saponaria officinalis Native Saporin (from Saponaria officinalis seeds)
Protéines et Peptides
Saporin is a ribosome-inactivating protein (RIP) of type I. This monomeric RNA N-glycosidase purified from seeds of the plant Saponaria officinalis also known as Soapwort, is capable of specific depurination of eukaryotic ribosomes thus arresting protein synthesis. No ligand has been identified in saporin hence its inability to transverse the cell membrane. Due to its toxicity and stability of the structure, saporin has proven extremely useful for construction of immunotoxins.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité