Anti-alpha synuclein Rabbit Polyclonal Antibody
Fournisseur: Biosensis
R-075-100
BSENR-075-100EA 302 EUR InStock
BSENR-075-100
Anti-alpha synuclein Rabbit Polyclonal Antibody
Anticorps
Alpha synuclein is an abundant 140 amino acid neuronal protein, expressed primarily at presynaptic terminals in the central nervous system. FUNCTION: May be involved in the regulation of dopamine release and transport. Soluble protein, normally localized primarily at the presynaptic region of axons, which can form filamentous aggregates that are the major non amyloid component of intracellular inclusions in several neurodegenerative diseases (synucleinopathies). Induces fibrillization of microtubule-associated protein tau. Reduces neuronal responsiveness to various apoptotic stimuli, leading to a decreased caspase 3 activation. TISSUE SPECIFICITY: Expressed principally in brain but is also expressed in low concentrations in all tissues examined except in liver. Concentrated in presynaptic nerve terminals.SUBUNIT: Soluble monomer which can form filamentous aggregates. Interacts with UCHL1. Interacts with phospholipase D and histones. SUBCELLULAR LOCATION: Cytoplasm. Membrane. Nucleus. Note=Membrane-bound in dopaminergic neurons. Also found in the nucleus. ALTERNATIVE PRODUCTS: 3 named isoforms produced by alternative splicing. Additional isoforms seem to exist.
Immunohistochemical and western blot analysis of human brain indicates a high level of specificity for this antiserum.
Application Information:
IHC, WB. A dilution of 1:500 to 1:3000 is recommended for both applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: alpha synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: alpha synuclein protein (aa 129-140)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité