Anti-Endomorphin-1 Rabbit Polyclonal Antibody
Fournisseur: Biosensis
R-1494-50
BSENR-1494-50EA 321 EUR InStock
BSENR-1494-50
Anti-Endomorphin-1 Rabbit Polyclonal Antibody
Anticorps
Endomorphins 1 and 2 are endogenous opioid peptides which have the highest affinity for the mu-opioid receptors. Located in various parts of the brain and interacts with mu-opioid receptors and produces analgesia.
Antibody recognizes endomorphin 1 by native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité