Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-TrkB (phospho Y816/Y817) Rabbit Polyclonal Antibody

Fournisseur: Biosensis
R-1717-50
BSENR-1717-50EA 302 EUR InStock
BSENR-1717-50
Anti-TrkB (phospho Y816/Y817) Rabbit Polyclonal Antibody
Anticorps
This antibody has been shown to be specific for TrkB phosphorylated on tyrosine 816/817 by phospho-peptide absorption dot blots, and on cell lysates from cell lines induced with retinoic acid and BDNF. Antibody detects a clear band in retinoic acid (RA) and BDNF-treated NSC34 cell lysates at ~120 kDa only, demonstrating that the phosphorylated TrkB receptor is being detected. While not fully tested, this antibody may detect phosphorylated TrkA (pY791/794, human/rodent) and TrkC (pY834/820/859, human/mouse/rat) due to high degree of amino acid homology surrounding the phosphorylation site.

This antibody reacts with human and mouse material. Other species not yet tested but it is predicted to react with rat and chicken TrkB-pY816/817 based on amino acid homology at the site.

Application information:
Western Blotting (0,5 to 2 µg/ml). High skim milk concentration (5%) causes suppression of pY817 signal, suggesting that 5% skim milk should be avoided as blocking buffer and antibody diluent. Strongest signal is obtained in 5% BSA blocking buffer, however, many non-specific bands are present. An equal mixture of skim milk and BSA (2,5% each) appears to provide the best compromise between signal and noise. However, optimization of blocking condition is recommended for each particular sample, with excess of BSA over skim milk likely to be beneficial for best results. Other applications have not been tested. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TrkB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Antigen location corresponds to 811AKASPVYLDILG822 in human TrkB. Phosphorylated Tyrosine target is amino acid number 817 in human, and 810AKASPVYLDILG821 in Uniprot# Q63604 (NTRK2_RAT) for rat and Uniprot# P15209 (NTRK2_MOUSE) for mouse. Phosphorylated Tyrosine target is amino acid number 816 in Rat and Mouse.
Host: Rabbit
Isotype:
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité